accessibility menu, dialog, popup

UserName

YYSSS

CAS RN 12039-19-9

IUPAC Name: diyttrium(3+) trisulfanediide
Synonyms: yttrium(III) sulfide
Molecular Weight (g/mol): 273.99
Molecular Formula: S3Y2
InChi Key: BPMRLDMEAPSVQN-UHFFFAOYSA-N
SMILES: [S--].[S--].[S--].[Y+3].[Y+3]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results