accessibility menu, dialog, popup

UserName

H2SiNbSiH2

CAS RN 12034-80-9

IUPAC Name: (silanylideneniobio)silane
Synonyms: (silanylideneniobio)silane
Molecular Weight (g/mol): 153.11
Molecular Formula: H4NbSi2
InChi Key: OVICFFSAIBBAPC-UHFFFAOYSA-N
SMILES: [SiH2]=[Nb]=[SiH2]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results