CAS RN 12033-62-4
CAS RN 12033-62-4
IUPAC Name:
tantalioamine
Synonyms:
tantalum nitride
Molecular Weight (g/mol):
194.96
Molecular Formula:
NTa
InChi Key:
MZLGASXMSKOWSE-UHFFFAOYSA-N
SMILES:
N#[Ta]
1
–
1
of
1
results
1
Tantalum nitride, 99.5% (metals basis)
CAS: 12033-62-4 Molecular Formula: NTa Molecular Weight (g/mol): 194.955 MDL Number: MFCD00049568 InChI Key: MZLGASXMSKOWSE-UHFFFAOYSA-N Synonym: tantalum nitride,tantalum nitride tan,tantalum mononitride,tantalum nitride trace metals basis 10g PubChem CID: 82832 IUPAC Name: azanylidynetantalum SMILES: N#[Ta]