accessibility menu, dialog, popup

UserName

MnMnSb

CAS RN 12032-97-2

IUPAC Name: antimony dimanganese
Synonyms: antimony dimanganese
Molecular Weight (g/mol): 231.64
Molecular Formula: Mn2Sb
InChi Key: CMWGISPXZSHYPK-UHFFFAOYSA-N
SMILES: [Mn].[Mn].[Sb]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results