accessibility menu, dialog, popup

UserName

CH3H3CCH3OCH3H3CH3CCH3POH3CH3CCH3OCH3CH3H3CCH3

CAS RN 1202865-21-1

IUPAC Name: 2-[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphanyl]benzaldehyde
Synonyms: 2-[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphanyl]benzaldehyde
Molecular Weight (g/mol): 574.79
Molecular Formula: C37H51O3P
InChi Key: PAWZHFURVJBYGV-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=C1C(C)(C)C)P(C1=CC(=C(OC)C(=C1)C(C)(C)C)C(C)(C)C)C1=CC=CC=C1C=O)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)