CAS RN 12023-53-9
IUPAC Name:
iron(3+) phosphanetriide
Synonyms:
iron phosphide
Molecular Weight (g/mol):
86.82
Molecular Formula:
FeP
InChi Key:
JNRQHASIGICJBU-UHFFFAOYSA-N
SMILES:
[P-3].[Fe+3]
1
–
1
of
1
results
1
Iron phosphide, 99.5% (metals basis)
CAS: 12023-53-9 Molecular Formula: Fe3P Molecular Weight (g/mol): 198.51 MDL Number: MFCD10567221 InChI Key: KFSKMQQNHSLZRJ-UHFFFAOYSA-N IUPAC Name: triferriophosphane SMILES: [Fe]P([Fe])[Fe]