CAS RN 120202-71-3
CAS RN 120202-71-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl (2R)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]acetate Hydrogen Sulfate ((R)-Clopidogrel Hydrogen Sulfate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.