CAS RN 1201944-91-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-((2,6-dichlorobenzyl)amino)-6,7-dimethoxyindeno[1,2-c]pyrazol-4(1H)-one, TRC
CAS: 1201944-91-3 Molecular Formula: C19H15Cl2N3O3 Molecular Weight (g/mol): 404.25 Synonym: [1,2-c]pyrazol-4(1H)-one, 3-[[(2,6-dichlorophenyl)methyl]amino]-6,7-dimethoxy-Indeno SMILES: COc1cc2C(=O)c3c(NCc4c(Cl)cccc4Cl)n[nH]c3c2cc1OC