CAS RN 120173-57-1
CAS RN 120173-57-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fmoc-Ser(O-α-D-GalNAc(OAc)3)-OH, MedChemExpress
MedChemExpress Fmoc-Ser(O-α-D-GalNAc(OAc)3)-OH is a drug for cancer.