accessibility menu, dialog, popup

UserName

CAS RN 12008-21-8

BBBBBBLa

CAS RN 12008-21-8

IUPAC Name: pentacyclo[3.1.0.0¹,³.0²,⁴.0⁴,⁶]hexaborane-1,4-diuide lanthanum
Synonyms: lanthanum hexaboride
Molecular Weight (g/mol): 203.77
Molecular Formula: B6La
InChi Key: IBDHMAYMLSYGAK-UHFFFAOYSA-N
SMILES: [La].B12B3[B-]11B4B1[B-]234
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results