CAS RN 1196509-60-0
CAS RN 1196509-60-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Velsecorat, MedChemExpress
MedChemExpress AZD7594 is a potent selective nonsteroidal glucocorticoid receptor modulator, with an IC50 of 0.9 nM.