CAS RN 1196-00-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
l-Isopinocampheol, TRC
CAS: 1196-00-5 Molecular Formula: C10 H18 O Molecular Weight (g/mol): 154.25 Synonym: Bicyclo[3.1.1]heptan-3-ol, 2,6,6-trimethyl-, (1R,2R,3R,5S)-,3-Pinanol, (1R,2R,3R,5S)-(-)- (8CI),Bicyclo[3.1.1]heptan-3-ol, 2,6,6-trimethyl-, [1R-(1α,2β,3α,5α)]-,(1R,2R,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptan-3-ol,(-)-Isopinocampheol,(1R,2R,3R,5S)-(-)-Isopinocampheol,(1R,2R,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptan-3-ol,l-Isopinocampheol IUPAC Name: (1S,3R,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptan-3-ol SMILES: C[C@H]1[C@H](O)C[C@@H]2C[C@H]1C2(C)C