CAS RN 119302-24-8
CAS RN 119302-24-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
(2Beta,3Alpha,5Alpha,16Beta,17Beta)-17-Acetoxy-3-hydroxy-2-(4-morpholinyl)-16-(1-pyrrolidinyl)andros, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3alpha-Hydroxy-2beta-(morpholin-4-yl)-16beta-(pyrrolidin-1-yl)-5alpha-androstan-17beta-yl Acetate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
2Beta,3Alpha,5Alpha,16Beta,17Beta)-17-Acetoxy-3-hydroxy-2-(4-morpholinyl)-16-(1-pyrrolidinyl)androstane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
(2Beta,3Alpha,5Alpha,16Beta,17Beta)-17-Acetoxy-3-hydroxy-2-(4-morpholinyl)-16-(1-pyrrolidinyl)androstane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.