CAS RN 119193-19-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(+)-(3R,4R)-3,4-Dimethyl-4-(3-hydroxyphenyl)piperidine, TRC
CAS: 119193-19-0 Molecular Formula: C13H19NO Molecular Weight (g/mol): 205.3 Synonym: 3-[(3R,4R)-3,4-Dimethyl-4-piperidinyl]phenol,(3R-cis)-3-(3,4-dimethyl-4-piperidinyl)phenol,(+)-(3R,4R)-3,4-Dimethyl-4-(3-hydroxyphenyl)piperidine,(+)-(3R,4R)-Dimethyl-4-(3-hydroxyphenyl)piperidine SMILES: C[C@H]1CNCC[C@@]1(C)c2cccc(O)c2