CAS RN 1187595-84-1
CAS RN 1187595-84-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Baricitinib phosphate, MedChemExpress
MedChemExpress Baricitinib phosphate (LY3009104 phosphate; INCB028050 phosphate) is a selective orally bioavailable JAK1/JAK2 inhibitor with IC50 of 5.9 nM and 5.7 nM, respectively.