CAS RN 1186426-66-3
CAS RN 1186426-66-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
DBPR108, MedChemExpress
MedChemExpress DBPR108 is a potent, selective, and orally bioavailable dipeptide-derived inhibitor of DPP4 with IC50 of 15 nM; no inhibition on DDP8 and DPP9.