CAS RN 1185108-16-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glutaric Acid-1,5-13C2, TRC
CAS: 1185108-16-0 Molecular Formula: C313C2H8O4 Molecular Weight (g/mol): 134.1 Synonym: Pentanedioic Acid-13C2,1,3-Propanedicarboxylic Acid-13C2,1,5-Pentanedioic Acid-13C2,NSC 9238-13C2 SMILES: O[13C](CCC[13C](O)=O)=O