CAS RN 1185071-13-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac 5-Hydroxymethyl Tolterodine-d14, TRC
CAS: 1185071-13-9 Molecular Formula: C22H17D14NO2 Molecular Weight (g/mol): 355.57 Synonym: 3-[3-[Bis(1-methylethyl)amino]-1-phenylpropyl]-4-hydroxybenzenemethanol SMILES: OC1=C(C(C2=CC=CC=C2)CCN(C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C(CO)C=C1