CAS RN 1184979-29-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Gimeracil-13C3, TRC
CAS: 1184979-29-0 Molecular Formula: C213C3H4ClNO2 Molecular Weight (g/mol): 148.52 Synonym: 5-Chloro-4-hydroxy-2(1H)-pyridinone-13C3,5-Chloro-2,4-dihydroxypyridine-13C3,5-Chloro-4-hydroxy-2-pyridone-13C3,Gimestat-13C3,CDHP -13C3 SMILES: O=C1C=[13C](O)[13C](Cl)=[13CH]N1