CAS RN 1175536-50-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2R,3R)- 2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)-2,3-butanediol Methanesulfonate, TRC
CAS: 1175536-50-1 Molecular Formula: C12H13F2N3O2 . CH 4O3S Molecular Weight (g/mol): 365.35 Synonym: (2R,3R)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butane-2,3-diol Methanesulfonate IUPAC Name: (2R,3R)-2-(2,4-difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butane-2,3-diol methanesulfonic acid SMILES: FC1=CC(F)=CC=C1[C@@]([C@H](O)C)(O)CN2C=NC=N2.CS(=O)(O)=O