accessibility menu, dialog, popup

UserName

CH3H3CCH3OONHOHO

CAS RN 117445-22-4

IUPAC Name: 3-({[(tert-butoxy)carbonyl]amino}methyl)benzoic acid
Synonyms: 3-{[(tert-butoxycarbonyl)amino]methyl}benzoic acid
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: MQNHKLMHRZYTBZ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)