CAS RN 1173018-74-0
CAS RN 1173018-74-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(±)-Promethazine-d4 HCl (phenothiazine-1,3,7,9-d4), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(±)-Promethazine-d4 HCl (phenothiazine-1,3,7,9-d4), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
3
(±)-Promethazine-d4 HCl (phenothiazine-1,3,7,9-d4), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.