CAS RN 1169870-80-7
IUPAC Name:
4-chloro-2-fluoro-3-methoxybenzoic acid
Synonyms:
4-chloro-2-fluoro-3-methoxybenzoic acid
Molecular Weight (g/mol):
204.58
Molecular Formula:
C8H6ClFO3
InChi Key:
ODAJPJWSLSBLLM-UHFFFAOYSA-N
SMILES:
COC1=C(Cl)C=CC(C(O)=O)=C1F
1
–
1
of
1
results
1
4-Chloro-2-fluoro-3-methoxybenzoic acid, 97%, Thermo Scientific™
CAS: 1169870-80-7 Molecular Formula: C8H6ClFO3 Molecular Weight (g/mol): 204.58 MDL Number: MFCD19687204 InChI Key: ODAJPJWSLSBLLM-UHFFFAOYSA-N PubChem CID: 57782418 IUPAC Name: 4-chloro-2-fluoro-3-methoxybenzoic acid SMILES: COC1=C(Cl)C=CC(C(O)=O)=C1F