accessibility menu, dialog, popup

UserName

NSOOClOHO

CAS RN 1164136-21-3

IUPAC Name: 1-(3-chlorobenzenesulfonyl)pyrrolidine-2-carboxylic acid
Synonyms: 1-(3-chlorobenzenesulfonyl)pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 289.73
Molecular Formula: C11H12ClNO4S
InChi Key: CXDCZBNJZWAXSZ-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCN1S(=O)(=O)C1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results