accessibility menu, dialog, popup

UserName

CAS RN 116313-85-0

OHOHONOO

CAS RN 116313-85-0

IUPAC Name: 3,4-dihydroxy-5-nitrobenzaldehyde
Synonyms: 3,4-dihydroxy-5-nitrobenzaldehyde
Molecular Weight (g/mol): 183.12
Molecular Formula: C7H5NO5
InChi Key: BBFJODMCHICIAA-UHFFFAOYSA-N
SMILES: OC1=CC(C=O)=CC(=C1O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results