CAS RN 1159-82-6
CAS RN 1159-82-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
10-Hydroxyamitriptyline, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
(5EZ,10RS)-5-[3-(Dimethyl-amino)propylidene]-10,11-dihydro-5H-dibenzo[a,d][7]annulen-10-ol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.