CAS RN 1158235-36-9
CAS RN 1158235-36-9
IUPAC Name:
4-(3-methoxyphenyl)pyrimidin-2-amine
Synonyms:
4-(3-methoxyphenyl)pyrimidin-2-amine
Molecular Weight (g/mol):
201.23
Molecular Formula:
C11H11N3O
InChi Key:
ZLLWVWJXTPKEQM-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(=C1)C1=NC(N)=NC=C1
1
–
1
of
1
results
1
2-Amino-4-(3-methoxyphenyl)pyrimidine, 95%, Thermo Scientific™
CAS: 1158235-36-9 Molecular Formula: C11H11N3O Molecular Weight (g/mol): 201.229 MDL Number: MFCD07381836 InChI Key: ZLLWVWJXTPKEQM-UHFFFAOYSA-N Synonym: 4-3-methoxyphenyl pyrimidin-2-amine,2-amino-4-3-methoxyphenyl pyrimidine,4-3-methoxyphenyl pyrimidine-2-ylamine,4-3-methoxyphenyl-2-pyrimidinamine,2-pyrimidinamine, 4-3-methoxyphenyl PubChem CID: 43839808 IUPAC Name: 4-(3-methoxyphenyl)pyrimidin-2-amine SMILES: COC1=CC=CC(=C1)C2=NC(=NC=C2)N