accessibility menu, dialog, popup

UserName

CAS RN 1150114-80-9

BNNH3COHOH

CAS RN 1150114-80-9

IUPAC Name: (1-methyl-1H-indazol-6-yl)boronic acid
Synonyms: 1-methylindazol-6-ylboronic acid
Molecular Weight (g/mol): 175.98
Molecular Formula: C8H9BN2O2
InChi Key: OMVQAWCHQBKZCF-UHFFFAOYSA-N
SMILES: CN1N=CC2=CC=C(C=C12)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results