accessibility menu, dialog, popup

UserName

CH3(R)OOHNH2

CAS RN 114926-39-5

IUPAC Name: 2-amino-3-(3-methylphenyl)propanoic acid
Synonyms: 3-methylphenylalanine
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: JZRBSTONIYRNRI-UHFFFAOYNA-N
SMILES: CC1=CC(CC(N)C(O)=O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)