accessibility menu, dialog, popup

UserName

CAS RN 1144573-61-4

IUPAC Name: potassium trifluoro(6-methoxypyridin-3-yl)boranuide
Synonyms: potassium trifluoro(6-methoxypyridin-3-yl)boranuide
Molecular Weight (g/mol): 215.02
Molecular Formula: C6H6BF3KNO
InChi Key: ASWXYOAUUZSFCK-UHFFFAOYSA-N
SMILES: [K+].COC1=NC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results