accessibility menu, dialog, popup

UserName

CAS RN 114-26-1

CH3HNOOOCH3H3C

CAS RN 114-26-1

IUPAC Name: 2-(propan-2-yloxy)phenyl N-methylcarbamate
Synonyms: rhoden boruho blattanex caswell no. 508 propoxylor tendex unden 50PM propoxur (substance) suncide bolfo
Molecular Weight (g/mol): 209.25
Molecular Formula: C11H15NO3
InChi Key: ISRUGXGCCGIOQO-UHFFFAOYSA-N
SMILES: CNC(=O)OC1=CC=CC=C1OC(C)C
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results