CAS RN 113507-82-7
CAS RN 113507-82-7
IUPAC Name:
1,1,2,2-tetrafluoro-2-(1,1,2,2,2-pentafluoroethoxy)ethane-1-sulfonic acid
Synonyms:
1,1,2,2-tetrafluoro-2-(1,1,2,2,2-pentafluoroethoxy)ethanesulfonic acid
Molecular Weight (g/mol):
316.09
Molecular Formula:
C4HF9O4S
InChi Key:
RJGUAQKOHAABLK-UHFFFAOYSA-N
SMILES:
OS(=O)(=O)C(F)(F)C(F)(F)OC(F)(F)C(F)(F)F
1
–
2
of
2
results
1
PFAS Mixture (PF4OPeA, PF5OHxA, 3,6-OPFHpA, PFEESA) 2 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Perfluoro(2-ethoxyethane) sulfonic acid 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality