CAS RN 113502-55-9
CAS RN 113502-55-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-Descarboxy Ketorolac, TRC
CAS: 113502-55-9 Molecular Formula: C14 H13 N O Molecular Weight (g/mol): 211.26 Synonym: Phenyl(2,3-dihydro-1H-pyrrolizin-5-yl)methanone,Ketorolac Trometamol Imp. I (EP) IUPAC Name: 6,7-dihydro-5H-pyrrolizin-3-yl(phenyl)methanone SMILES: O=C(c1ccccc1)c2ccc3CCCn23
2
Phenyl(2,3-dihydro-1H-pyrrolizin-5-yl)methanone, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.