accessibility menu, dialog, popup

UserName

H3CSNH2OO

CAS RN 1135-00-8

IUPAC Name: 4-butylbenzene-1-sulfonamide
Synonyms: P-butylbenzenesulfonamide
Molecular Weight (g/mol): 213.3
Molecular Formula: C10H15NO2S
InChi Key: IWEMSJAOIKQBBZ-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)S(N)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results