CAS RN 112776-37-1
CAS RN 112776-37-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Benzylidene-3,4-(methylenedioxy)benzylamine, 98+%, Thermo Scientific™
CAS: 112776-37-1 Molecular Formula: C15H13NO2 MDL Number: MFCD00115461