CAS RN 1126320-68-0
IUPAC Name:
4-chloro-2-fluoro-3-methoxybenzonitrile
Synonyms:
4-chloro-2-fluoro-3-methoxybenzonitrile
Molecular Weight (g/mol):
185.58
Molecular Formula:
C8H5ClFNO
InChi Key:
UKAFKFNOOLUFSX-UHFFFAOYSA-N
SMILES:
COC1=C(Cl)C=CC(C#N)=C1F
1
–
1
of
1
results
1
4-Chloro-2-fluoro-3-methoxybenzonitrile, 97%, Thermo Scientific™
CAS: 1126320-68-0 Molecular Formula: C8H5ClFNO Molecular Weight (g/mol): 185.58 MDL Number: MFCD19687208 InChI Key: UKAFKFNOOLUFSX-UHFFFAOYSA-N PubChem CID: 59126941 IUPAC Name: 4-chloro-2-fluoro-3-methoxybenzonitrile SMILES: COC1=C(Cl)C=CC(C#N)=C1F