CAS RN 1105698-15-4
IUPAC Name:
N-(3-{[(2-oxo-1,2-dihydronaphthalen-1-ylidene)methyl]amino}phenyl)-2-phenylpropanamide
Synonyms:
N-(3-{[(2-oxonaphthalen-1-ylidene)methyl]amino}phenyl)-2-phenylpropanamide
Molecular Weight (g/mol):
394.47
Molecular Formula:
C26H22N2O2
InChi Key:
UADRPWLRVLBVBC-UHFFFAOYNA-N
SMILES:
CC(C(=O)NC1=CC=CC(NC=C2C(=O)C=CC3=CC=CC=C23)=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Salermide, MedChemExpress
MedChemExpress Salermide is an inhibitor of Sirt1 and Sirt2; can cause strong cancer-specific apoptotic cell death.