CAS RN 109304-69-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
2-(Acetylamino)-2-deoxy-4,6-O-(phenylmethylene)-Alpha-D-glucopyranose 1-[Bis(phenylmethyl) Phosphate], TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(Acetylamino)-2-deoxy-4,6-O-(phenylmethylene)-Alpha-D-glucopyranose 1-[Bis(phenylmethyl) Phosphate], TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(Acetylamino)-2-deoxy-4,6-O-(phenylmethylene)-a-D-glucopyranose 1-[Bis(phenylmethyl) Phosphate], TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.