CAS RN 1092813-99-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lisinopril (8R,S)-Diketopiperazine (Mixture of Diastereomers)Mps1-IN-1 dihydrochloride, TRC
CAS: 1092813-99-4 Molecular Formula: C21 H29 N3 O4 Molecular Weight (g/mol): 387.47 Synonym: Lisinopril (8R,S)-Diketopiperazine IUPAC Name: (2S)-2-[(3S)-3-(4-aminobutyl)-1,4-dioxo-6,7,8,8a-tetrahydro-3H-pyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoic acid SMILES: NCCCC[C@@H]1N([C@@H](CCc2ccccc2)C(=O)O)C(=O)C3CCCN3C1=O