accessibility menu, dialog, popup

UserName

CAS RN 1092460-86-0

IUPAC Name: [5-fluoro-2-(trifluoromethoxy)phenyl]methanol
Synonyms: [5-fluoro-2-(trifluoromethoxy)phenyl]methanol
Molecular Weight (g/mol): 210.13
Molecular Formula: C8H6F4O2
InChi Key: JKXVLACUMFQMNS-UHFFFAOYSA-N
SMILES: OCC1=C(OC(F)(F)F)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results