accessibility menu, dialog, popup

UserName

HClH3CNCH3Cl

CAS RN 109-54-6

IUPAC Name: hydrogen (3-chloropropyl)dimethylamine chloride
Synonyms: hydrogen (3-chloropropyl)dimethylamine chloride
Molecular Weight (g/mol): 158.07
Molecular Formula: C5H13Cl2N
InChi Key: LJQNMDZRCXJETK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCCCl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results