CAS RN 1083065-04-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Amino-5,7-dihydro-5-[(4-methoxyphenyl)methyl]-6H-dibenz[b,d]azepin-6-one Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.