CAS RN 108149-61-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
3-(1,1-Dimethylethyl) 4-methyl (4S,5R)-2,2,5-trimethyl-3,4-oxazolidinedicarboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-(1,1-Dimethylethyl) 4-methyl (4S,5R)-2,2,5-trimethyl-3,4-oxazolidinedicarboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-(1,1-Dimethylethyl) 4-methyl (4S,5R)-2,2,5-trimethyl-3,4-oxazolidinedicarboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.