CAS RN 108-09-8
IUPAC Name:
4-methylpentan-2-amine
Synonyms:
2-pentanamine, 4-methyl-
Molecular Weight (g/mol):
101.19
Molecular Formula:
C6H15N
InChi Key:
UNBMPKNTYKDYCG-UHFFFAOYNA-N
SMILES:
CC(C)CC(C)N
1
–
2
of
2
results
1
1,3-Dimethylbutylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,3-Dimethylbutylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.