CAS RN 107952-99-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(11Beta)-11,21-Bis(acetyloxy)pregna-4,16-diene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(11Beta)-11,21-Bis(acetyloxy)pregna-4,16-diene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.