CAS RN 1079-21-6
IUPAC Name:
[1,1'-biphenyl]-2,5-diol
Synonyms:
phenylhydroquinone
Molecular Weight (g/mol):
186.21
Molecular Formula:
C12H10O2
InChi Key:
XCZKKZXWDBOGPA-UHFFFAOYSA-N
SMILES:
OC1=CC=C(O)C(=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Phenylhydroquinone, TRC
CAS: 1079-21-6 Molecular Formula: C12 H10 O2 Molecular Weight (g/mol): 186.21 Synonym: Phenyl-2,5-biphenyldioll Hydroquinone,Phenyl-1,4-benzenediol,1,4-Dihydroxy-2-phenylbenzene,2,5-Dihydroxybiphenyl,2-Phenyl-1,4-benzenediol,2-Phenyl-1,4-dihydroxybenzene,2-Phenyl-1,4-hydroquinone,2-Phenylhydroquinone,NSC 407988,Phenyl-p-hydroquinone,Phenylhydroquinone IUPAC Name: 2-phenylbenzene-1,4-diol SMILES: Oc1ccc(O)c(c1)c2ccccc2