CAS RN 107864-90-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(aR)-rel-a-((4-Chlorophenyl)-a-(1-cyclopropylethyl)-1H-1,2,4-triazole-1-ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.