CAS RN 107658-27-5
IUPAC Name:
pyridin-3-yl trifluoromethanesulfonate
Synonyms:
pyridin-3-yl trifluoromethanesulfonate
Molecular Weight (g/mol):
227.16
Molecular Formula:
C6H4F3NO3S
InChi Key:
ZRNDSLLIAOLRJS-UHFFFAOYSA-N
SMILES:
FC(F)(F)S(=O)(=O)OC1=CN=CC=C1
1
–
3
of
3
results
1
3-Pyridyl Trifluoromethanesulfonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Pyridyl Trifluoromethanesulfonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Pyridyl Trifluoromethanesulfonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.