CAS RN 1076199-83-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-6-[N,N-di(2-hydroxybenzyl)amino]-9-isopropylpurine, TRC
CAS: 1076199-83-1 Molecular Formula: C22 H22 Cl N5 O2 Molecular Weight (g/mol): 423.9 IUPAC Name: 2-[[(2-chloro-9-propan-2-ylpurin-6-yl)-[(2-hydroxyphenyl)methyl]amino]methyl]phenol SMILES: CC(C)n1cnc2c(nc(Cl)nc12)N(Cc3ccccc3O)Cc4ccccc4O