CAS RN 1076198-22-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
4-(2-Hydroxy-1,1-dimethylethyl)-N-[6-chloro-5-(2-methoxyphenoxy)-(2,2'-bipyrimidin)-4-yl]benzenesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(2-Hydroxy-1,1-dimethylethyl)-N-[6-chloro-5-(2-methoxyphenoxy)-(2,2'-bipyrimidin)-4-yl]benzenesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(2-Hydroxy-1,1-dimethylethyl)-N-[6-chloro-5-(2-methoxyphenoxy)-(2,2'-bipyrimidin)-4-yl]benzenesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.